4,4'-Biphenyldiboronic acid bis(pinacol)ester 97%
|
|
BD236676-10G
BD236676-250MG
BLDPBD236676-10GEA
102
EUR
BLDPBD236676-10G
BLDPBD236676-250MG
4,4'-Biphenyldiboronic acid bis(pinacol)ester 97%
4,4'-Biphenyldiboronic acid bis(pinacol) ester
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₂₄H₃₂B₂O₄ Storage Temperature: Refrigerator |
MDL Number:
MFCD08669545 CAS Number: 207611-87-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...