2-(Piperidin-1-ylmethyl)phenylboronic acid 98%

BD236738-1G BD236738-5G
BLDPBD236738-1GEA 185 EUR
BLDPBD236738-1G BLDPBD236738-5G
2-(Piperidin-1-ylmethyl)phenylboronic acid 98%
2-(Piperidin-1-ylmethyl)phenylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₂H₁₈BNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD06801661
CAS Number: 878289-33-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR