3-(Methylsulphonyl)propyl 4-methylbenzenesulphonate 98%

BD236818-5G BD236818-250MG
BLDPBD236818-5GEA 558 EUR
BLDPBD236818-5G BLDPBD236818-250MG
3-(Methylsulphonyl)propyl 4-methylbenzenesulphonate 98%
3-(Methylsulphonyl)propyl 4-methylbenzenesulphonate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₁H₁₆O₅S₂
MW: 292.37 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22124592
CAS Number: 263400-88-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR