2,5-Dichloro-N-[2-(isopropylsulphonyl)phenyl]pyrimidin-4-amine 98%

BD237827-5G BD237827-1G
BLDPBD237827-5GEA 92.4 EUR
BLDPBD237827-5G BLDPBD237827-1G
2,5-Dichloro-N-[2-(isopropylsulphonyl)phenyl]pyrimidin-4-amine 98%
2,5-Dichloro-N-[2-(isopropylsulphonyl)phenyl]pyrimidin-4-amine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₁₃H₁₃Cl₂N₃O₂S
Storage Temperature: Ambient
MDL Number: MFCD12827956
CAS Number: 761440-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR