Thiomorpholine-1,1-dioxide 95%

BD2384-5G BD2384-25G
BLDPBD2384-5GEA 17.9 EUR
BLDPBD2384-5G BLDPBD2384-25G
Thiomorpholine-1,1-dioxide 95%
Thiomorpholine 1,1-dioxide

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₄H₉NO₂S
Storage Temperature: Ambient
MDL Number: MFCD05861623
CAS Number: 39093-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR