(1R,2R)-N1,N1-Dimethylcyclohexane-1,2-diamine 99+%

BD238589-1G BD238589-100MG
BLDPBD238589-1GEA 81.9 EUR
BLDPBD238589-1G BLDPBD238589-100MG
(1R,2R)-N1,N1-Dimethylcyclohexane-1,2-diamine 99+%
(1R,2R)-N1,N1-Dimethylcyclohexane-1,2-diamine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₈H₁₈N₂
Storage Temperature: Ambient
MDL Number: MFCD09054929
CAS Number: 320778-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR