(S)-3-Aminohexan-1-ol hydrochloride 97%
|
|
BD238614-5G
BD238614-1G
BLDPBD238614-5GEA
588
EUR
BLDPBD238614-5G
BLDPBD238614-1G
(S)-3-Aminohexan-1-ol hydrochloride 97%
(S)-3-Aminohexan-1-ol hydrochloride
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₆H₁₆ClNO Storage Temperature: Ambient |
MDL Number:
MFCD23701471 CAS Number: 68889-62-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...