2,5-Dibromo-3,4-pyridinediamine 98%

BD238859-10G BD238859-5G
BLDPBD238859-10GEA 431 EUR
BLDPBD238859-10G BLDPBD238859-5G
2,5-Dibromo-3,4-pyridinediamine 98%
2,5-Dibromo-3,4-pyridinediamine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₅H₅Br₂N₃
Storage Temperature: Refrigerator
MDL Number: MFCD15526814
CAS Number: 221241-11-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR