5-Chloropyrimidine-4,6-diol 95%

BD239071-100MG BD239071-1G
BLDPBD239071-100MGEA 11.6 EUR
BLDPBD239071-100MG BLDPBD239071-1G
5-Chloropyrimidine-4,6-diol 95%
5-Chloropyrimidine-4,6-diol

Storage condition: Inert atmosphere, 2-8°C

Formula: C₄H₃ClN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD18374057
CAS Number: 1193-56-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR