4-Aminotetrahydro-2H-pyran acetate 98%

BD239102-5G BD239102-25G BD239102-1G
BLDPBD239102-5GEA 33.6 EUR
BLDPBD239102-5G BLDPBD239102-25G BLDPBD239102-1G
4-Aminotetrahydro-2H-pyran acetate 98%
4-Aminotetrahydro-2H-pyran acetate

Storage condition: Inert atmosphere, 2-8°C

Formula: C₅H₁₁NO·CH₃CO₂H
Storage Temperature: Refrigerator
MDL Number: MFCD08445669
CAS Number: 1005498-91-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR