Methyl-3-amino-4-nitrobenzoate 97%
|
|
BD239333-100MG
BD239333-1G
BLDPBD239333-100MGEA
27.3
EUR
BLDPBD239333-100MG
BLDPBD239333-1G
Methyl-3-amino-4-nitrobenzoate 97%
Methyl-3-amino-4-nitrobenzoate
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formula:
C₈H₈N₂O₄ Storage Temperature: Ambient |
MDL Number:
MFCD00017008 CAS Number: 99512-09-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...