Methyl 2-(2-fluoro-4-hydroxyphenyl)acetate 95%

BD239363-100MG BD239363-25G
BLDPBD239363-100MGEA 7.4 EUR
BLDPBD239363-100MG BLDPBD239363-25G
Methyl 2-(2-fluoro-4-hydroxyphenyl)acetate 95%
Methyl 2-(2-fluoro-4-hydroxyphenyl)acetate

Storage condition: Inert atmosphere, Room Temperature

Formula: C₉H₉FO₃
Storage Temperature: Ambient
MDL Number: MFCD18399286
CAS Number: 91361-59-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR