Methyl 4-amino-3-(methylamino)benzoate 98%

BD239583-250MG BD239583-1G
BLDPBD239583-250MGEA 53.6 EUR
BLDPBD239583-250MG BLDPBD239583-1G
Methyl 4-amino-3-(methylamino)benzoate 98%
Methyl 4-amino-3-(methylamino)benzoate

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₉H₁₂N₂O₂
MW: 180.20 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD15144544
CAS Number: 616224-38-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR