(1-Ethyl-1H-imidazol-5-yl)methanol 95% (contains < 3% H₂O)
|
|
BD240827-1G
BD240827-250MG
BLDPBD240827-1GEA
164
EUR
BLDPBD240827-1G
BLDPBD240827-250MG
(1-Ethyl-1H-imidazol-5-yl)methanol 95% (contains < 3% H₂O)
(1-Ethyl-1H-imidazol-5-yl)methanol
Storage condition: Sealed in dry, Room Temperature
Formula:
C₆H₁₀N₂O Storage Temperature: Ambient |
MDL Number:
MFCD04114647 CAS Number: 215872-62-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...