Piperazin-2-one, N1-BOC protected 95%

BD241151-5G BD241151-250MG
BLDPBD241151-5GEA 767 EUR
BLDPBD241151-5G BLDPBD241151-250MG
Piperazin-2-one, N1-BOC protected 95%
Piperazin-2-one, N1-BOC protected

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₉H₁₆N₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD06797597
CAS Number: 889958-14-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR