Ethyl-1H-pyrrolo[2,3-b]pyridine-4-carboxylate 95%
BD241484-250MG
BD241484-1G
BLDPBD241484-250MGEA
122
EUR
BLDPBD241484-250MG
BLDPBD241484-1G
Ethyl-1H-pyrrolo[2,3-b]pyridine-4-carboxylate 95%
Ethyl-1H-pyrrolo[2,3-b]pyridine-4-carboxylate
Storage condition: Sealed in dry, Room Temperature
Formula:
C₁₀H₁₀N₂O₂ |
MDL Number:
MFCD15071133 CAS Number: 1261588-72-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...