2-Chloro-5-methoxy-4-pyrimidinamine 97%

BD242255-25G BD242255-10G
BLDPBD242255-25GEA 576 EUR
BLDPBD242255-25G BLDPBD242255-10G
2-Chloro-5-methoxy-4-pyrimidinamine 97%
2-Chloro-5-methoxy-4-pyrimidinamine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₅H₆ClN₃O
Storage Temperature: Refrigerator
MDL Number: MFCD17169913
CAS Number: 99979-77-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR