1,1-Bis(hydroxymethyl)cyclopropane 98%

BD2438-500G BD2438-5G
BLDPBD2438-500GEA 116 EUR
BLDPBD2438-500G BLDPBD2438-5G
1,1-Bis(hydroxymethyl)cyclopropane 98%
1,1-Bis(hydroxymethyl)cyclopropane

Storage condition: Sealed in dry, Room Temperature

Formula: C₅H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD00464207
CAS Number: 39590-81-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR