(R)-2-(((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)methyl)-3-methylbutanoic acid 98%
|
|
BD244650-250MG
BD244650-5G
BLDPBD244650-250MGEA
57.8
EUR
BLDPBD244650-250MG
BLDPBD244650-5G
(R)-2-(((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)methyl)-3-methylbutanoic acid 98%
(R)-2-(((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)methyl)-3-methylbutanoic acid
Storage condition: Sealed in dry, 2-8°C
Formula:
C₂₁H₂₃NO₄ Storage Temperature: Refrigerator |
MDL Number:
MFCD07372882 CAS Number: 501331-02-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...