2'-Chloroacetanilide 98%

BD244976-25G BD244976-10G
BLDPBD244976-25GEA 24.2 EUR
BLDPBD244976-25G BLDPBD244976-10G
2'-Chloroacetanilide 98%
2'-Chloroacetanilide

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₈ClNO
Storage Temperature: Ambient
MDL Number: MFCD00045169
CAS Number: 533-17-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR