Disodium hexahydroxoplatinate 98%

BD246752-250MG BD246752-100MG
BLDPBD246752-250MGEA 72.5 EUR
BLDPBD246752-250MG BLDPBD246752-100MG
Disodium hexahydroxoplatinate 98%
Disodium hexahydroxoplatinate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: H₆Na₂O₆Pt
MW: 343.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00014236
CAS Number: 12325-31-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR