3-(Methylamino)-1-(thiophen-2-yl)propan-1-one hydrochloride 97%
|
|
BD247251-5G
BD247251-10G
BLDPBD247251-5GEA
58.8
EUR
BLDPBD247251-5G
BLDPBD247251-10G
3-(Methylamino)-1-(thiophen-2-yl)propan-1-one hydrochloride 97%
3-(Methylamino)-1-(thiophen-2-yl)propan-1-one hydrochloride
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formula:
C₈H₁₂ClNOS MW: 205.71 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD23703103 CAS Number: 645411-16-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...