Methyl 2,4-dichloro-6-methylpyrimidine-5-carboxylate 95%

BD247376-25G BD247376-1G
BLDPBD247376-25GEA 2360 EUR
BLDPBD247376-25G BLDPBD247376-1G
Methyl 2,4-dichloro-6-methylpyrimidine-5-carboxylate 95%
Methyl 2,4-dichloro-6-methylpyrimidine-5-carboxylate

Storage condition: Inert atmosphere, 2-8°C

Formula: C₇H₆Cl₂N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD22572319
CAS Number: 36745-93-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR