4,7-Bis(5-bromo-2-thienyl)-2,1,3-benzothiadiazole 95%
|
|
BD247404-5G
BD247404-1G
BLDPBD247404-5GEA
745
EUR
BLDPBD247404-5G
BLDPBD247404-1G
4,7-Bis(5-bromo-2-thienyl)-2,1,3-benzothiadiazole 95%
4,7-Bis(5-bromo-2-thienyl)-2,1,3-benzothiadiazole
Storage condition: Keep in dark place, Inert atmosphere, 2-8°C
Formula:
C₁₄H₆Br₂N₂S₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD16619295 CAS Number: 288071-87-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...