2,6-Dibromo-5-chloronicotinic acid 95%
|
|
BD248726-250MG
BD248726-100MG
BLDPBD248726-250MGEA
156
EUR
BLDPBD248726-250MG
BLDPBD248726-100MG
2,6-Dibromo-5-chloronicotinic acid 95%
2,6-Dibromo-5-chloronicotinic acid
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₆H₂Br₂ClNO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD26743442 CAS Number: 55304-86-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...