3-Hydroxy-1-methylazetidine hydrochloride 98%

BD250538-1G BD250538-10G
BLDPBD250538-1GEA 81.9 EUR
BLDPBD250538-1G BLDPBD250538-10G
3-Hydroxy-1-methylazetidine hydrochloride 98%
3-Hydroxy-1-methylazetidine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₄H₉NO·ClH
Storage Temperature: Ambient
MDL Number: MFCD16605978
CAS Number: 26687-49-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR