4-Bromo-5-methoxybenzene-1,2-diamine 95%

BD250733-100MG BD250733-1G
BLDPBD250733-100MGEA 114 EUR
BLDPBD250733-100MG BLDPBD250733-1G
4-Bromo-5-methoxybenzene-1,2-diamine 95%
4-Bromo-5-methoxybenzene-1,2-diamine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₇H₉BrN₂O
Storage Temperature: Refrigerator
MDL Number: MFCD21648471
CAS Number: 108447-01-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR