Ethyl 4-[(3-chloro-4-methoxybenzyl)amino]-2-(methylthio)pyrimidine-5-carboxylate 98%

BD250967-25G BD250967-1G
BLDPBD250967-25GEA 129 EUR
BLDPBD250967-25G BLDPBD250967-1G
Ethyl 4-[(3-chloro-4-methoxybenzyl)amino]-2-(methylthio)pyrimidine-5-carboxylate 98%
Ethyl 4-[(3-chloro-4-methoxybenzyl)amino]-2-(methylthio)pyrimidine-5-carboxylate

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Formula: C₁₆H₁₈ClN₃O₃S
Storage Temperature: Freezer
MDL Number: MFCD17012695
CAS Number: 330785-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR