(E)-3-(m-Tolyl)acrylaldehyde 95%
|
|
BD251568-100G
BD251568-250MG
BLDPBD251568-100GEA
1240
EUR
BLDPBD251568-100G
BLDPBD251568-250MG
(E)-3-(m-Tolyl)acrylaldehyde 95%
(E)-3-(m-Tolyl)acrylaldehyde
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₁₀H₁₀O Storage Temperature: Refrigerator |
MDL Number:
MFCD08460293 CAS Number: 93614-80-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...