Dimethyl-trans-cyclohexane-1,2-dicarboxylate 96%

BD251906-5G BD251906-250MG
BLDPBD251906-5GEA 344 EUR
BLDPBD251906-5G BLDPBD251906-250MG
Dimethyl-trans-cyclohexane-1,2-dicarboxylate 96%
Dimethyl trans-cyclohexane-1,2-dicarboxylate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₁₆O₄
Storage Temperature: Ambient
MDL Number: MFCD09998658
CAS Number: 3205-35-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR