N-Fmoc-D-aspartic acid-4-allyl ester 95%

BD252020-25G BD252020-10G
BLDPBD252020-25GEA 415 EUR
BLDPBD252020-25G BLDPBD252020-10G
N-Fmoc-D-aspartic acid-4-allyl ester 95%
N-Fmoc-D-aspartic acid-4-allyl ester

Storage condition: Sealed in dry, 2-8°C

Formula: C₂₂H₂₁NO₆
Storage Temperature: Refrigerator
MDL Number: MFCD01074688
CAS Number: 177609-12-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR