4-Ethynylpiperidine hydrochloride 95%

BD253116-5G BD253116-1G
BLDPBD253116-5GEA 516 EUR
BLDPBD253116-5G BLDPBD253116-1G
4-Ethynylpiperidine hydrochloride 95%
4-Ethynylpiperidine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₇H₁₂ClN
Storage Temperature: Ambient
MDL Number: MFCD18910999
CAS Number: 550378-30-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR