2,4-Dichloropyridine 98%
|
|
BD2535-10G
BD2535-5G
BLDPBD2535-10GEA
11.6
EUR
BLDPBD2535-10G
BLDPBD2535-5G
2,4-Dichloropyridine 98%
2,4-Dichloropyridine
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₅H₃Cl₂N Storage Temperature: Ambient |
MDL Number:
MFCD00955618 CAS Number: 26452-80-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...