6-Chloro-2-fluoropyridin-3-ol 98%

BD254794-5G BD254794-1G BD254794-10G
BLDPBD254794-5GEA 490 EUR
BLDPBD254794-5G BLDPBD254794-1G BLDPBD254794-10G
6-Chloro-2-fluoropyridin-3-ol 98%
6-Chloro-2-fluoropyridin-3-ol

Storage condition: Inert atmosphere, Room Temperature

Formula: C₅H₃ClFNO
Storage Temperature: Ambient
MDL Number: MFCD07437887
CAS Number: 883107-68-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR