5-bromo-4-methoxypyrimidine 98%

BD254928-250MG BD254928-25G
BLDPBD254928-250MGEA 57.8 EUR
BLDPBD254928-250MG BLDPBD254928-25G
5-bromo-4-methoxypyrimidine 98%
5-bromo-4-methoxypyrimidine

Storage condition: Sealed in dry, Room Temperature

Formula: C₅H₅BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD00234066
CAS Number: 4319-85-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR