(R)-1-(3-(Trifluoromethyl)phenyl)ethanamine hydrochloride 95%

BD254991-10G BD254991-100MG
BLDPBD254991-10GEA 1020 EUR
BLDPBD254991-10G BLDPBD254991-100MG
(R)-1-(3-(Trifluoromethyl)phenyl)ethanamine hydrochloride 95%
(R)-1-(3-(Trifluoromethyl)phenyl)ethanamine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₉H₁₁ClF₃N
Storage Temperature: Ambient
MDL Number: MFCD11100947
CAS Number: 1213630-93-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR