Oxetan-3-yl methanesulphonate 95%

BD255761-5G BD255761-250MG
BLDPBD255761-5GEA 450 EUR
BLDPBD255761-5G BLDPBD255761-250MG
Oxetan-3-yl methanesulphonate 95%
Oxetan-3-yl methanesulphonate

Storage condition: Inert atmosphere, 2-8°C

Formula: C₄H₈O₄S
MW: 152.169 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18207677
CAS Number: 148430-81-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR