2-(4-Bromo-3-chlorophenyl)acetonitrile 96%

BD256297-1G BD256297-100MG
BLDPBD256297-1GEA 619 EUR
BLDPBD256297-1G BLDPBD256297-100MG
2-(4-Bromo-3-chlorophenyl)acetonitrile 96%
2-(4-Bromo-3-chlorophenyl)acetonitrile

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₅BrClN
Storage Temperature: Ambient
MDL Number: MFCD18390590
CAS Number: 1259023-29-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR