4,7-Dihydroimidazo[4,5-d][1,3]diazepin-8(1H)-one hydrochloride 95%

BD256745-250MG BD256745-50MG
BLDPBD256745-250MGEA 402 EUR
BLDPBD256745-250MG BLDPBD256745-50MG
4,7-Dihydroimidazo[4,5-d][1,3]diazepin-8(1H)-one hydrochloride 95%
4,7-Dihydroimidazo[4,5-d][1,3]diazepin-8(1H)-one hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₆H₇ClN₄O
Storage Temperature: Ambient
MDL Number: MFCD23105840
CAS Number: 71222-44-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR