4-(3,4-Dichlorophenoxy)piperidine hydrochloride 98%
|
|
BD25704-1G
BD25704-250MG
BLDPBD25704-1GEA
325
EUR
BLDPBD25704-1G
BLDPBD25704-250MG
4-(3,4-Dichlorophenoxy)piperidine hydrochloride 98%
4-(3,4-Dichlorophenoxy)piperidine hydrochloride
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₁₁H₁₄Cl₃NO Storage Temperature: Ambient |
MDL Number:
MFCD03840126 CAS Number: 817186-93-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...