(2R,3S)-3-(tert-Butoxy)-2-(4-ethyl-2,3-dioxopiperazine-1-carboxamido)butanoic acid 97%

BD257137-1G BD257137-25G
BLDPBD257137-1GEA 8.4 EUR
BLDPBD257137-1G BLDPBD257137-25G
(2R,3S)-3-(tert-Butoxy)-2-(4-ethyl-2,3-dioxopiperazine-1-carboxamido)butanoic acid 97%
(2R,3S)-3-(tert-Butoxy)-2-(4-ethyl-2,3-dioxopiperazine-1-carboxamido)butanoic acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₅H₂₅N₃O₆
Storage Temperature: Ambient
MDL Number: MFCD29050420
CAS Number: 79276-23-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR