2,6-Dichloro-3-pyridineboronic acid 97%

BD2578-1G BD2578-25G
BLDPBD2578-1GEA 11.6 EUR
BLDPBD2578-1G BLDPBD2578-25G
2,6-Dichloro-3-pyridineboronic acid 97%
2,6-Dichloro-3-pyridineboronic acid

Storage condition: Inert atmosphere, 2-8°C

Formula: C₅H₄BCl₂NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD03094989
CAS Number: 148493-34-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR