2-Bromo-6-(trifluoromethyl)benzonitrile 95%

BD257982-250MG BD257982-5G
BLDPBD257982-250MGEA 45.2 EUR
BLDPBD257982-250MG BLDPBD257982-5G
2-Bromo-6-(trifluoromethyl)benzonitrile 95%
2-Bromo-6-(trifluoromethyl)benzonitrile

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₃BrF₃N
MW: 250.0153 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08741382
CAS Number: 914637-35-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR