2-Amino-3-bromo-5-(trifluoromethoxy)benzoic acid 98%

BD258000-1G BD258000-250MG
BLDPBD258000-1GEA 71.4 EUR
BLDPBD258000-1G BLDPBD258000-250MG
2-Amino-3-bromo-5-(trifluoromethoxy)benzoic acid 98%
2-Amino-3-bromo-5-(trifluoromethoxy)benzoic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₈H₅BrF₃NO₃
MW: 300.029 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03407438
CAS Number: 874774-41-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR