1-Bromo-2-chloro-3,5-difluorobenzene 98%
|
|
BD258049-5G
BD258049-10G
BLDPBD258049-5GEA
83
EUR
BLDPBD258049-5G
BLDPBD258049-10G
1-Bromo-2-chloro-3,5-difluorobenzene 98%
1-Bromo-2-chloro-3,5-difluorobenzene
Storage condition: Sealed in dry, Room Temperature
Formula:
C₆H₂BrClF₂ Storage Temperature: Ambient |
MDL Number:
MFCD00135441 CAS Number: 187929-82-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...