7-Methoxyindoline hydrochloride 95+%
|
|
BD259327-250MG
BD259327-100MG
BLDPBD259327-250MGEA
56.7
EUR
BLDPBD259327-250MG
BLDPBD259327-100MG
7-Methoxyindoline hydrochloride 95+%
7-Methoxyindoline hydrochloride
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₉H₁₂ClNO Storage Temperature: Ambient |
MDL Number:
MFCD11506101 CAS Number: 4770-43-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...