4-Amino-3,5-dichloropyridine 98%
|
|
BD2595-100G
BD2595-5G
BLDPBD2595-100GEA
65.1
EUR
BLDPBD2595-100G
BLDPBD2595-5G
4-Amino-3,5-dichloropyridine 98%
4-Amino-3,5-dichloropyridine
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formula:
C₅H₄Cl₂N₂ Storage Temperature: Ambient |
MDL Number:
MFCD00125023 CAS Number: 22889-78-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...