Methyl 4-chloro-1H-indole-6-carboxylate 95%
|
|
BD261378-250MG
BD261378-100MG
BLDPBD261378-250MGEA
124
EUR
BLDPBD261378-250MG
BLDPBD261378-100MG
Methyl 4-chloro-1H-indole-6-carboxylate 95%
Methyl 4-chloro-1H-indole-6-carboxylate
Storage condition: Sealed in dry, Room Temperature
Formula:
C₁₀H₈ClNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD07781543 CAS Number: 885522-78-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...