3-Oxo-3,4-dihydropyrazine-2-carboxylic acid 97%
|
|
BD2614-100G
BD2614-5G
BLDPBD2614-100GEA
226
EUR
BLDPBD2614-100G
BLDPBD2614-5G
3-Oxo-3,4-dihydropyrazine-2-carboxylic acid 97%
3-Oxo-3,4-dihydropyrazine-2-carboxylic acid
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formula:
C5H4N2O3 Storage Temperature: Ambient |
MDL Number:
MFCD00235136 CAS Number: 20737-42-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...