Methyl-5-bromo-1H-indole-3-carboxylate 98%

BD26166-10G BD26166-5G
BLDPBD26166-10GEA 152 EUR
BLDPBD26166-10G BLDPBD26166-5G
Methyl-5-bromo-1H-indole-3-carboxylate 98%
Methyl-5-bromo-1H-indole-3-carboxylate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₈BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD06203728
CAS Number: 773873-77-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR